missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Bifluoride, Purified, Reagents

Catalog No. 01337506
Change view
Click to view available options
Quantity:
100 mL
500 g
3 kg
10 kg
25 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337506 100 mL
01337505 10 kg
01337504 25 kg
01337503 3 kg
01337502 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01337506 Supplier Labchem Inc Supplier No. C2130500100A6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ammonium Bifluoride, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1341-49-7
Molecular Formula F2H5N
Molecular Weight (g/mol) 57.04
MDL Number MFCD00012018
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
Synonym Ammonium Hydrogen Difluoride, Ammonium Fluoride Hydrofluoride
IUPAC Name amine dihydrofluoride
SMILES N.F.F

Specifications

Boiling Point 240°C
Melting Point 124.6°C
Color Colorless/White
Physical Form Solid
Quantity 100 mL
Grade Pure
Packaging Poly Bottle
Specific Gravity 1.5
Chemical Name or Material Ammonium Bifluoride, Purified