missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Bifluoride, Flake, 95%, Spectrum™ Chemical
Specifications
Specifications
| Physical Form | Flakes or Powder |
| CAS Max % | 1 |
| Quantity | 25 g |
| Packaging | Poly Pail |
| Molecular Weight (g/mol) | 57.05 |
| Percent Purity | 95% |
| Grade | Reagent |
| Chemical Name or Material | Ammonium Bifluoride |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.