missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Bifluoride (Crystalline/Technical), Fisher Chemical™

Catalog No. p-30850
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A664500 500 g Poly Bottle
A6643 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A664500 Supplier Thermo Fisher Scientific Supplier No. FLA664500
Only null left

Chemical Identifiers

CAS 1341-49-7
Molecular Formula F2H5N
Molecular Weight (g/mol) 57.04
MDL Number MFCD00012018
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
Synonym Ammonium Acid Fluoride, Ammonium Hydrogen Difluoride
IUPAC Name amine dihydrofluoride
SMILES N.F.F

Specifications

Boiling Point 230°C
Melting Point 125°C
Color White
pH 3.5
Physical Form Solid
Quantity 500 g
Formula Weight 57.04
Grade Technical
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 1hPa at 20°C
Decomposition Information >230°C
Density 1.50g/cm³
Chemical Name or Material Ammonium Bifluoride
Show More Show Less
DOTInformation DOT Class 8, : Corrosive