missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium bicarbonate, Puriss., meets analytical spec. of Ph.Eur., BP, E 503, 99-101%, Honeywell Fluka™
Puriss., meets analytical specification of Ph.Eur., BP, E 503, 99-101%
$285.50 - $285.50
Chemical Identifiers
| CAS | 1066-33-7 |
|---|---|
| Molecular Formula | CH5NO3 |
| Molecular Weight (g/mol) | 79.06 |
| MDL Number | MFCD00012138 |
| InChI Key | ATRRKUHOCOJYRX-UHFFFAOYSA-N |
| Synonym | ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327 |
| PubChem CID | 14013 |
| IUPAC Name | carbonic acid amine |
| SMILES | N.OC(O)=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 1066-33-7 | |
| 79.06 | |
| ATRRKUHOCOJYRX-UHFFFAOYSA-N | |
| 14013 | |
| N.OC(O)=O |
| CH5NO3 | |
| MFCD00012138 | |
| ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327 | |
| carbonic acid amine |
Specifications
| 1066-33-7 | |
| CH5NO3 | |
| MFCD00012138 | |
| ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327 | |
| N.OC(O)=O | |
| 79.06 | |
| 79.06g/mol | |
| puriss. | |
| Vapor Density: 2.7 (vs air) |
| 1 kg | |
| NH4HCO3 | |
| 4329606 | |
| ATRRKUHOCOJYRX-UHFFFAOYSA-N | |
| carbonic acid amine | |
| 14013 | |
| 99 to 101% | |
| Poly bottle | |
| Ammonium Bicarbonate, Meets analytical specification of Ph.Eur.BP |
Safety and Handling
P301 + P312 + P330
H302
EINECSNumber : 213-911-5
RTECSNumber : BO8600000