missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Bicarbonate (Powder/Certified), Fisher Chemical™

Catalog No. p-30842
Change view
Click to view available options
Quantity:
500 g
50 kg
Packaging:
Fiber Drum
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A643500 500 g Poly Bottle
A64350 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A643500 Supplier Thermo Fisher Scientific Supplier No. FLA643500
Only null left

Chemical Identifiers

CAS 1066-33-7
Molecular Formula CH5NO3
Molecular Weight (g/mol) 79.06
MDL Number MFCD00012138
InChI Key ATRRKUHOCOJYRX-UHFFFAOYSA-N
Synonym ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
PubChem CID 14013
IUPAC Name carbonic acid amine
SMILES N.OC(O)=O

Specifications

Melting Point 60°C
Color White
pH 7.8
Physical Form Solid
Quantity 500 g
Assay Percent Range 21.30 to 21.73 %
Linear Formula NH4HCO3
Formula Weight 79.06
Percent Purity 21.30 to 21.73%
Grade Certified
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 60mmHg at 25°C
Density 1.586g/cm³
Chemical Name or Material Ammonium Bicarbonate
Show More Show Less