missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Bicarbonate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. p-5390677
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Catalog No. Quantity Packaging
02002270 500 g Poly Bottle
02002269 2.5 kg Poly Bottle
2 options

Catalog No. 02002270

Supplier: Avantor 300301

Only null left

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 1066-33-7
Molecular Formula CH5NO3
Molecular Weight (g/mol) 79.06
MDL Number MFCD00012138
InChI Key ATRRKUHOCOJYRX-UHFFFAOYSA-N
Synonym ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
PubChem CID 14013
IUPAC Name carbonic acid amine
SMILES N.OC(O)=O

Specifications

Physical Form Powder
Quantity 500 g
Formula Weight 79.06
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.59kg
Chemical Name or Material Ammonium Bicarbonate