missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Bicarbonate Crystalline MP Biomedicals
Supplier: MP Biomedicals Inc 0215010790
Specifications
| Ammonium Bicarbonate | |
| Crystalline | |
| CH5NO3 | |
| ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327 | |
| N.OC(O)=O | |
| 79.06 | |
| 79.1 |
| 1066-33-7 | |
| 500 g | |
| MFCD00012138 | |
| ATRRKUHOCOJYRX-UHFFFAOYSA-N | |
| carbonic acid amine | |
| 14013 | |
| 1.586 g/cm3 |
Chemical Identifiers
| 1066-33-7 | |
| 79.06 | |
| ATRRKUHOCOJYRX-UHFFFAOYSA-N | |
| 14013 | |
| N.OC(O)=O |
| CH5NO3 | |
| MFCD00012138 | |
| ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327 | |
| carbonic acid amine |