missing translation for 'onlineSavingsMsg'
Learn More

Ammonium bicarbonate, 99%, for analysis

CAS: 1066-33-7 | C3H8N2O9Zn2 | 346.86 g/mol

Supplier:  Thermo Scientific Chemicals 393210010

 View more versions of this product

Catalog No. AC393210010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonium bicarbonate
1066-33-7
Crystalline Powder
99%
NH4HCO3
15, 494
Solubility in water: 220g/L (20°C)
N.OC(O)=O
79.06
79.06
Analytical
105.0°C
White
1 kg
CH5NO3
MFCD00012138
ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
ATRRKUHOCOJYRX-UHFFFAOYSA-N
carbonic acid amine
14013
99%
Plastic bottle

Chemical Identifiers

1066-33-7
79.06
ATRRKUHOCOJYRX-UHFFFAOYSA-N
14013
N.OC(O)=O
CH5NO3
MFCD00012138
ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
carbonic acid amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.

GHS Signal Word: Warning

EINECSNumber : 213-911-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only