missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Alum, Dodecahydrate, USP, 99-100.5%, Spectrum™ Chemical

Catalog No. p-7156955
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600561 500 g Poly Bottle
18600562 2.5 kg Poly Pail
18600563 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18600561 Supplier Spectrum Chemical Mfg Cor Supplier No. A1621500GM
Only null left

A1621, 7784-26-1, AlNH4(SO4)2·12H2O

Spectrum™ Chemical Ammonium Alum, Dodecahydrate, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7784-26-1
Molecular Formula AlH28NO20S2
Molecular Weight (g/mol) 453.31
InChI Key WZUKKIPWIPZMAS-UHFFFAOYSA-K
IUPAC Name aluminum(3+) ammonium dodecahydrate disulfate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 99 to 100.5%
Grade USP
Loss on Drying 45.0-48%
Packaging Poly Bottle