missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Alum, Dodecahydrate, USP, 99-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Ammonium Alum, Dodecahydrate, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Physical Form | Crystalline Powder or Needles |
| Quantity | 500 g |
| Percent Purity | 99 to 100.5% |
| Grade | USP |
| Loss on Drying | 45.0-48% |
| Packaging | Poly Bottle |
| Chemical Name or Material | Ammonium Alum, Dodecahydrate |