missing translation for 'onlineSavingsMsg'
Learn More

Ammonium acetate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥98%, Honeywell Fluka™

Catalog No. 60046128
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 500 g
10 kg
20 kg
6 x 1 kg
6 x 2.5 kg
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60046128 6 x 500 g
6002405 100 g
6002410 500 g
6002407 1 kg
6002408 2.5 kg
60046126 6 x 1 kg
6002406 10 kg
60046127 6 x 2.5 kg
6002409 20 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 60046128 Supplier Honeywell Chemicals Supplier No. 323016X500G
Only null left

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Melting Point 110°C to 112°C (decomposition)(literature)
CAS Max % ≤100.0000%
Quantity 6 x 500 g
Linear Formula CH3CO2NH4
Beilstein 4186741
Formula Weight 77.08
Percent Purity >98%
Grade ACS Reagent/Reagent Ph. Eur.
Packaging Poly Bottle
Density 1.170 g/cm3
Chemical Name or Material Ammonium acetate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥98%
Show More Show Less
ShelfLife 1620 days from date of manufacture
EINECSNumber 211-162-9
Recommended Storage Room Temp