missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate, Purified, Spectrum™ Chemical

$249.53 - $6270.87

Chemical Identifiers

CAS 631-61-8
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18608820
View Documents
Spectrum Chemical Mfg Cor
A112125GM
25 g Poly Bottle
Each for $249.53
Only null left
 
18608818
View Documents
Spectrum Chemical Mfg Cor
A1121500GM
500 g Poly Bottle
Each for $381.53
Only null left
 
18608819
View Documents
Spectrum Chemical Mfg Cor
A112125KG
2.5 kg Poly Bottle
Each for $1,284.93
Only null left
 
18608816
View Documents
Spectrum Chemical Mfg Cor
A112112KG
12 kg Poly Pail
Each for $2,942.67
Only null left
 
18608817
View Documents
Spectrum Chemical Mfg Cor
A112145KGBL
45 kg Fiber Drum
Each for $6,270.87
Only null left
 
Description

Description

Specifications

Chemical Identifiers

631-61-8

Specifications

631-61-8
25 g
Poly Bottle
1
Purified
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.