missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate, LC-MS Ultra, Honeywell™

Catalog No. 6002013
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002013 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6002013 Supplier Honeywell Chemicals Supplier No. 1426725G
Only null left

CHROMASOLV™ LC-MS Ultra; eluent additive for UHPLC-MS.

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise.
  • Exemplary lot-to-lot reproducibility
  • Low levels of trace metals
  • Packaged in plastic bottles

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Quantity 25 g
Linear Formula CH3CO2NH4
Beilstein 4186741
Formula Weight 77.08g/mol
Grade LC-MS Ultra
Packaging Plastic Bottles
Residue after Ignition ≤0.01% (as SO4)
Chemical Name or Material Ammonium Acetate
EINECSNumber 211-162-9
RTECSNumber AF3675000