missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Acetate, HPLC Grade, J.T. Baker™
Description
- For Use in High Performance Liquid Chromatography; For Laboratory,Research,or Manufacturing Use
Specifications
Specifications
| Quantity | 1 kg |
| Formula Weight | 77.08 |
| Percent Purity | ≥97% |
| Grade | HPLC |
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 1.07kg |
| Chemical Name or Material | Ammonium Acetate |