missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Acetate, HPLC Grade, 97%, Spectrum™ Chemical
A2149, 631-61-8, C2H7NO2
$367.23 - $1162.60
Chemical Identifiers
| CAS | 631-61-8 |
|---|---|
| Molecular Formula | C2H7NO2 |
| Molecular Weight (g/mol) | 77.08 |
| InChI Key | USFZMSVCRYTOJT-UHFFFAOYSA-N |
| IUPAC Name | ammonium acetate |
| SMILES | [NH4+].CC([O-])=O |