missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate, Honeywell

Catalog No. 6002704
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002704 250 g
6002705 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002704 Supplier Honeywell Chemicals Supplier No. 431311250G
Only null left

≥99.99% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Quantity 250 g
Linear Formula CH3CO2NH4
Beilstein 4186741
Formula Weight 77.08g/mol
Percent Purity ≥99.99% (trace metals basis)
Chemical Name or Material Ammonium acetate
RTECSNumber AF3675000