Learn More
Ammonium Acetate (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical FLA63710
Specifications
| Ammonium Acetate | |
| 631-61-8 | |
| 7.0 | |
| 10 kg | |
| C2H7NO2 | |
| MFCD00013066 | |
| USFZMSVCRYTOJT-UHFFFAOYSA-N | |
| azanium;acetate | |
| 517165 | |
| 77.08 | |
| Certified ACS | |
| Poly Pail |
| 114°C | |
| White | |
| Solid | |
| ≥97 % | |
| CH3COONH4 | |
| ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
| CC(=O)[O-].[NH4+] | |
| 77.083 | |
| CHEBI:62947 | |
| ≥97% | |
| Pass Test |
Chemical Identifiers
| 631-61-8 | |
| 77.083 | |
| USFZMSVCRYTOJT-UHFFFAOYSA-N | |
| 517165 | |
| azanium;acetate |
| C2H7NO2 | |
| MFCD00013066 | |
| ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
| CHEBI:62947 | |
| CC(=O)[O-].[NH4+] |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:1
Instability:1