missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. p-5390676
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Poly Bottle
Catalog No. Quantity Packaging
02002268 500 g Poly Bottle
02002267 2 kg Poly Bottle
2 options

Catalog No. 02002268

Supplier: Avantor 059601

Only null left

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Physical Form Crystals
Quantity 500 g
Formula Weight 77.08
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.07kg
Chemical Name or Material Ammonium Acetate