missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate Buffer, pH 5, Reagents

Catalog No. 01336160
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336160 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336160 Supplier Labchem Inc Supplier No. CA0008001A

Ammonium Acetate Buffer, pH 5, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Buffer
CAS 7732-18-5
Color Colorless
pH 5
Physical Form Liquid
Quantity 1 L
Packaging Poly Bottle
Sterility Non-sterile
Solubility Information Completely miscible