missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium acetate, 97+%, MP Biomedicals™
$91.96 - $136.00
Chemical Identifiers
| CAS | 631-61-8 |
|---|---|
| Molecular Formula | C2H7NO2 |
| Molecular Weight (g/mol) | 77.083 |
| InChI Key | USFZMSVCRYTOJT-UHFFFAOYSA-N |
| Synonym | ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 |
| PubChem CID | 517165 |
| ChEBI | CHEBI:62947 |
| IUPAC Name | azanium;acetate |
| SMILES | CC(=O)[O-].[NH4+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19140480
|
MP Biomedicals Inc
0219140480 |
100 g |
Each for $91.96
|
|
|||||
|
ICN19140490
|
MP Biomedicals Inc
0219140490 |
500 g |
Each for $136.00
|
|
|||||
Chemical Identifiers
| 631-61-8 | |
| 77.083 | |
| ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
| CHEBI:62947 | |
| CC(=O)[O-].[NH4+] |
| C2H7NO2 | |
| USFZMSVCRYTOJT-UHFFFAOYSA-N | |
| 517165 | |
| azanium;acetate |
Specifications
| White | |
| Powder or Liquid | |
| 0.97+ | |
| ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
| CC(=O)[O-].[NH4+] | |
| 77.083 | |
| CHEBI:62947 | |
| 1.17 (water = 1) |
| 631-61-8 | |
| 100 g | |
| C2H7NO2 | |
| USFZMSVCRYTOJT-UHFFFAOYSA-N | |
| azanium;acetate | |
| 517165 | |
| 77.08 | |
| Ammonium acetate |