missing translation for 'onlineSavingsMsg'
Learn More

Ammonium acetate, 97+%, MP Biomedicals™

Numéro de catalogue. ICN19140490
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
ICN19140490 500 g
ICN19140480 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. ICN19140490 Fournisseur MP Biomedicals Inc Code fournisseur 0219140490
Il en reste null

Identifiants chimiques

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Spécifications

Color White
Physical Form Powder or Liquid
Quantity 500 g
Assay Percent Range 0.97+
Formula Weight 77.08
Density 1.17 (water = 1)
Chemical Name or Material Ammonium acetate