missing translation for 'onlineSavingsMsg'
Learn More

Ammonium acetate, ≥99.0% (calc. on dry substance, T), MilliporeSigma™ Supelco™

Catalog No. p-7188015
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018829 25 g
111018073 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111018829 Supplier Merck Emd Millipore Supplier No. 7359425GF
Only null left

Ammonium acetate exists as a white crystal. It is prepared from acetic acid and ammonia by neutralization to pH 7.0. It is widely used in analytical chemistry, drugs and in textile dyeing.

Chemical Identifiers

MDL Number MFCD00013066

Specifications

pH 6.2 to 7.5 (25°C, 1 M aq. soln.)
Melting Point 110°C to 112°C
CAS 631-61-8
Linear Formula CH3 CO2 NH4
Solubility Information Water: 1 M (at 20°C, clear, colorless)
Formula Weight 77.08
Percent Purity ≥99.0% (calc. on dry substance, T)
Grade Mass Spec
Quantity 25 g
Physical Form Solid
RTECSNumber AF3675000