missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate, ∽98%, MP Biomedicals

$97.56 - $132.13

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19400080
View Documents
MP Biomedicals Inc
0219400080
100 g
Each for $97.56
Only null left
 
ICN19400090
View Documents
MP Biomedicals Inc
0219400090
500 g
Each for $132.13
Only null left
 
Description

Description

Specifications

Chemical Identifiers

631-61-8
77.083
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
CHEBI:62947
CC(=O)[O-].[NH4+]
C2H7NO2
USFZMSVCRYTOJT-UHFFFAOYSA-N
517165
azanium;acetate

Specifications

631-61-8
∽98%
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
CC(=O)[O-].[NH4+]
77.083
CHEBI:62947
Ammonium Acetate
100 g
C2H7NO2
USFZMSVCRYTOJT-UHFFFAOYSA-N
azanium;acetate
517165
77.1
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'