missing translation for 'onlineSavingsMsg'
Learn More

Ammonium acetate, 5M aq. soln.

Catalog No. p-7041911
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60688AD 50 mL
AAJ60688AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60688AD Supplier Thermo Scientific Chemicals Supplier No. J60688AD
Only null left

CAS: 631-61-8 | C2H7NO2 | 77.083 g/mol

Ammonium acetate is used for routine precipitation of nucleic acids, and is especially useful for reducing the coprecipitation of unwanted dNTPs and contaminating oligosaccharides. It is used as an important reagent for studying molecular biology, biological buffers, reagents and DNA and RNA purification. It is a popular buffer for mobile phases for HPLC with ESLD detection, for ESI mass spectrometry of proteins and other molecules, and has been used to replace cell buffers with non-volitile salts. Ammonium acetate is also used in protein studies and protein preparation. It can be used in the protein purification steps of dialysis to remove contaminants through diffusion and, when combined with distilled water, as a protein precipitating agent. In organic chemistry, ammonium acetate solution is useful as a catalyst in the Knoevenagel condensation and as a source of ammonia in the Borch reaction. Additionally, it is occasionally used commercially as a biodegradable de-icing agent and as an additive in food as an acidity regulator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium acetate is used for routine precipitation of nucleic acids, and is especially useful for reducing the coprecipitation of unwanted dNTPs and contaminating oligosaccharides. It is used as an important reagent for studying molecular biology, biological buffers, reagents and DNA and RNA purification. It is a popular buffer for mobile phases for HPLC with ESLD detection, for ESI mass spectrometry of proteins and other molecules, and has been used to replace cell buffers with non-volitile salts. Ammonium acetate is also used in protein studies and protein preparation. It can be used in the protein purification steps of dialysis to remove contaminants through diffusion and, when combined with distilled water, as a protein precipitating agent. In organic chemistry, ammonium acetate solution is useful as a catalyst in the Knoevenagel condensation and as a source of ammonia in the Borch reaction. Additionally, it is occasionally used commercially as a biodegradable de-icing agent and as an additive in food as an acidity regulator.

Notes
Store only at -20°C in a freezer without a defrost cycle.

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 50 mL
Linear Formula CH3COONH4
Solubility Information Soluble in alcohol (freely),acetone (slightly),water (148g/100ml) at 4°C,and water (7.89g/100ml) at 15°C.
Formula Weight 77.08
Concentration or Composition (by Analyte or Components) 5M aq. soln.
Chemical Name or Material Ammonium acetate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only