missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium 1-Pyrrolidinecarbodithioate 95.0+%, TCI America™
Supplier: TCI America P0644250G
Specifications
| Ammonium 1-Pyrrolidinecarbodithioate [Reagent for Atomic Absorption Analysis] | |
| 144°C | |
| C5H8NS2- | |
| 250 g | |
| VSWDORGPIHIGNW-UHFFFAOYSA-M | |
| pyrrolidine-1-carbodithioate | |
| 4311638 | |
| ≥95.0% (T) |
| 5108-96-3 | |
| White-Yellow | |
| MFCD00012720 | |
| ammonium pyrrolidyldithiocarbamate, apdc, ammonium 1-pyrrolidinecarbodithioate, ammonium tetramethylenedithiocarbamate, ammonium pyrrolidine-1-carbodithioate, ammonium 1-pyrrolidinyldithiocarbamate, ammonium 1-pyrrolidinedithiocarboxylate, 1-pyrrolidinecarbodithioic acid, ammonium salt, 1-pyrrolidinecarbodithioic acid ammonium salt, pyrrolidinedithiocarbamate ammonium | |
| C1CCN(C1)C(=S)[S-] | |
| 146.246 | |
| 164.29 | |
| Crystalline Powder |
Chemical Identifiers
| 5108-96-3 | |
| 146.246 | |
| VSWDORGPIHIGNW-UHFFFAOYSA-M | |
| 4311638 | |
| C1CCN(C1)C(=S)[S-] |
| C5H8NS2- | |
| MFCD00012720 | |
| ammonium pyrrolidyldithiocarbamate, apdc, ammonium 1-pyrrolidinecarbodithioate, ammonium tetramethylenedithiocarbamate, ammonium pyrrolidine-1-carbodithioate, ammonium 1-pyrrolidinyldithiocarbamate, ammonium 1-pyrrolidinedithiocarboxylate, 1-pyrrolidinecarbodithioic acid, ammonium salt, 1-pyrrolidinecarbodithioic acid ammonium salt, pyrrolidinedithiocarbamate ammonium | |
| pyrrolidine-1-carbodithioate |
Safety and Handling
EINECSNumber : (5)-0111
TSCA : Yes
Recommended Storage : Freezer