missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia Solution, FCC, 27-30%, Spectrumâ„¢ Chemical
A1293, 1336-21-6, NH4OH
$516.90 - $9133.63
Chemical Identifiers
CAS | 1336-21-6 |
---|---|
Molecular Formula | H5NO |
Molecular Weight (g/mol) | 35.05 |
MDL Number | MFCD00066650 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
IUPAC Name | amine hydrate |
SMILES | N.O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
18600546
|
Spectrum Chemical Mfg Cor
A1293500MLGL |
500 mL | Amber Glass Bottle |
Each for $516.90
|
|
|||||
18600541
|
Spectrum Chemical Mfg Cor
A129325LTPV |
2.5 L | Poly-Coated Glass Bottle |
Each for $671.97
|
|
|||||
18600547
|
Spectrum Chemical Mfg Cor
A1293500MLGLCS6 |
6 x 500 mL | Amber Glass Bottle |
Case of 6 Each for $1,690.37
|
|
|||||
18600542
|
Spectrum Chemical Mfg Cor
A12934LTPV |
4 L | Poly-Coated Glass Bottle |
Each for $882.10
|
|
|||||
18600543
|
Spectrum Chemical Mfg Cor
A129325LTPVCS4 |
4 x 2.5 L | Poly-Coated Glass Bottle |
Case of 4 Each for $1,779.47
|
|
|||||
18600544
|
Spectrum Chemical Mfg Cor
A12934LTPVCS4 |
4 x 4 L | Poly-Coated Glass Bottle |
Case of 4 Each for $2,283.07
|
|
|||||
18600545
|
Spectrum Chemical Mfg Cor
A129320LT |
20 L | Poly Drum |
Each for $2,737.27
|
|
|||||
18600548
|
Spectrum Chemical Mfg Cor
A1293200LTBL |
200 L | Drum |
Each for $9,133.63
|
|
|||||
Description
Spectrumâ„¢ Chemical Ammonia Solution, FCC is a solution of Ammonia in water. It has many uses, but in food production it is used primarily as a leavening agent. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
1336-21-6 | |
35.05 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
N.O |
H5NO | |
MFCD00066650 | |
amine hydrate |
Specifications
1336-21-6 | |
H5NO | |
UN2672 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
amine hydrate | |
27 to 30% | |
Amber Glass Bottle |
100% | |
MFCD00066650 | |
500 mL | |
N.O | |
35.05 | |
FCC |