missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ammonia Reagent

Catalog No. TR60101
Change view
Click to view available options
Quantity:
2 x 28 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TR60101 2 x 28 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TR60101 Supplier Thermo Scientific™ Supplier No. JTR60101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Determine quantity of ammonia in plasma.

Determine quantity of ammonia in plasma in vitro with Thermo Scientific™ Ammonia Reagent.

Excess ammonia exerts toxic effects on the central nervous system. Elevated levels of ammonia may be either due to inborn errors of metabolism or secondary to other conditions. Elevated ammonia may be observed in advanced liver disease caused by Reye's Syndrome, viral hepatitis or cirrhosis.

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
Description Ammonia Reagent
CE Marker Yes
Detectable Analytes Ammonia
Sample Type Plasma
Type Reagent
Linearity 0 to 1000µmol/L
Methods Compatibility Kinetic
Quantity 2 x 28 mL
Stability Stable Liquid
State at Room Temperature Liquid
Form Liquid
Show More Show Less