missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Ammonia (IIAR) Pipe Markers, Legend: HTRL/LIQ/Ammonia/LOW

For use in labeling ammonia piping and its contents. Brady Ammonia (IIAR) Pipe Markers help create a safer, more efficient work environment.

$42.27

Specifications

Color Black, White, Orange, Green, Yellow
Product Type Ammonia (IIAR) Pipe Marker
Display Text IIAR Abbreviation: HTRL; Physical State: LIQ; Pressure Level: LOW; Legend: Ammonia
For Use With (Application) Pipe and valve marking
View More Specs

Products 1
Catalog Number Mfr. No. Description Material Diameter (English) Price Quantity & Availability  
Catalog Number Mfr. No. Description Material Diameter (English) Price Quantity & Availability  
19110517
View Documents Promotion Details
Brady™
59928
Style HPHV Polyester Over-laminate (B-681), Fiberglass Plastic Carrier (B-883) 8 in.
Each for $42.27
Only null left
 
Description

Description

  • Meets IIAR Standard and is recommended for use in labeling ammonia piping and its contents
  • Made of B-689 high performance wrap-around PVF over-laminated polyester and B-681/B-883 polyester over-laminate on fiberglass plastic carrier
Specifications

Specifications

Black, White, Orange, Green, Yellow
Ammonia (IIAR) Pipe Marker
IIAR Abbreviation: HTRL; Physical State: LIQ; Pressure Level: LOW; Legend: Ammonia
Pipe and valve marking
Videos
SDS
Documents

Documents