missing translation for 'onlineSavingsMsg'
Learn More

Ammonia, ca. 7N solution in methanol

Catalog No. p-3734403
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Plastic Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC133710250 25 mL Glass Bottle
AC133710010 1 L Glass Bottle
AC133710025 2.5 L Glass Bottle
AC133710251 25 L Plastic Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC133710250 Supplier Thermo Scientific Chemicals Supplier No. 133710250
Only null left

Ammonia, H3N, CAS Number-7664-41-7, 67-56-1, am-fol, ammonia, ammonia anhydrous, ammonia, anhydrous, ammonia gas, ammoniak, ammoniakgas, anhydrous ammonia, nitro-sil, spirit of hartshorn, 2.5L, CHEBI:16134, Colorless to Yellow, 6.0 to 7.5N, 0.7700g/mL, 05,15, 14 deg.C, 17.03, GHS Signal Word: Danger | CAS: 7664-41-7 | H3N | 17.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Ammonia
Color Colorless to Yellow
Name Note ca. 7N Solution in Methanol
CAS 7664-41-7, 67-56-1
Molecular Formula H3N
Linear Formula NH3
Concentration 6.0 to 7.5N
MDL Number MFCD00011418
Fieser 05,15
Merck Index 15, 488
Density 0.7700g/mL
Synonym ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
Quantity 25 mL
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
SMILES N
IUPAC Name azane
Molecular Weight (g/mol) 17.03
PubChem CID 222
Flash Point 14°C
ChEBI CHEBI:16134
Formula Weight 17.03
Packaging Glass Bottle
Specific Gravity 0.77
Physical Form Liquid
Show More Show Less

RUO – Research Use Only