missing translation for 'onlineSavingsMsg'
Learn More

Ammeline 95.0+%, TCI America™

Catalog No. p-7116999
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A06765G 5 g
A067625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A06765G Supplier TCI America Supplier No. A06765G
Only null left

Chemical Identifiers

CAS 645-92-1
Molecular Formula C3H5N5O
Molecular Weight (g/mol) 127.107
MDL Number MFCD00023186
InChI Key MASBWURJQFFLOO-UHFFFAOYSA-N
Synonym ammeline, 4,6-diamino-1,3,5-triazin-2-ol, ammelin, 1,3,5-triazin-2 1h-one, 4,6-diamino, 2,4-diamino-6-hydroxy-1,3,5-triazine, cyanurodiamide, 2,4-diamino-1,3,5-triazin-6-one, 4,6-diamino-1,3,5-triazin-2 1h-one, unii-lc8g9556q0, s-triazin-2-ol, 4,6-diamino
PubChem CID 12583
ChEBI CHEBI:28646
IUPAC Name 2,6-diamino-1H-1,3,5-triazin-4-one
SMILES C1(=NC(=O)N=C(N1)N)N

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 127.11
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Ammeline
EINECSNumber (5)-5439
TSCA Yes