missing translation for 'onlineSavingsMsg'
Learn More

Amlodipine Related Compound D, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111022995
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022995 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022995 Supplier Merck Emd Millipore Supplier No. PHR205350MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

Quantity 50 mg
Molecular Formula C28H27ClN2O7
Linear Formula C28H27ClN2O7
Synonym 3-Ethyl 5-methyl (4 RS)-4-(2-chlorophenyl)-2-[[2-(1,3-dioxo-1,3-dihydro-2 H-isoindol-2-yl)ethoxy]methyl]-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate; 3-Ethyl 5-methyl 4-(2-chlorophenyl)-2-{[2-(1,3-dioxoisoindolin-2-yl)ethoxy]methyl}-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate; Phthalimidoamlodipine
Formula Weight 538.98
Grade Reference Material