missing translation for 'onlineSavingsMsg'
Learn More

Amlodipine Besylate 98.0+%, TCI America™

Catalog No. A23531G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23531G 1 g
A23535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23531G Supplier TCI America Supplier No. A23531G

Chemical Identifiers

CAS 111470-99-6
Molecular Formula C26H31ClN2O8S
Molecular Weight (g/mol) 567.05
MDL Number MFCD00887594
InChI Key ZPBWCRDSRKPIDG-UHFFFAOYSA-N
Synonym amlodipine besylate, amlodipine besilate, cardiorex, amlodipine benzenesulfonate, istin, amdepin, amdipin, amlodin, amlogard, amlopin
PubChem CID 60496
ChEBI CHEBI:2669
IUPAC Name benzenesulfonic acid;3-O-ethyl 5-O-methyl 2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C(C1C2=CC=CC=C2Cl)C(=O)OC)C)COCCN.C1=CC=C(C=C1)S(=O)(=O)O

Specifications

Melting Point 200°C
Color Green-White
Quantity 1 g
Formula Weight 567.05
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Amlodipine Besylate
RTECSNumber US7967700
TSCA No