missing translation for 'onlineSavingsMsg'
Learn More

Amlodipine, 97+%

Catalog No. AAJ6224209
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6224209 10 g
AAJ6224203 1 g
AAJ6224206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6224209 Supplier Thermo Scientific Chemicals Supplier No. J6224209

A calcium channel antagonist with potent antioxidant activity | CAS: 88150-42-9 | C20H25ClN2O5 | 408.88 g/mol

A calcium channel antagonist with potent antioxidant activityAmlodipine is an L-type calcium channel protein inhibitor and blocker. It is used in combination with the angiotensin receptor blocker olmesartan. Further, it acts as a dihydropyridine calcium channel blocker. It is used as an anti-hypertensive. It is involved in the treatment of angina and to lower blood pressure. In addition to this, it is used to prevent chest pain and in coronary artery disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethyl sulfoxide, ethanol, water, dimethyl formamide, acetone, chloroform, ethyl acetate and methanol.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 88150-42-9
Molecular Formula C20H25ClN2O5
Molecular Weight (g/mol) 408.88
MDL Number MFCD00864687
InChI Key HTIQEAQVCYTUBX-UHFFFAOYNA-N
Synonym amlodipine, norvasc, amlodis, amlodipino, amlodipinum, amlocard, coroval, lipinox, amlor, amlodipinum latin
PubChem CID 2162
ChEBI CHEBI:2668
IUPAC Name 3-ethyl 5-methyl 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(COCCN)NC(C)=C(C1C1=CC=CC=C1Cl)C(=O)OC

Specifications

Melting Point 134°C to 136°C
Color White
Quantity 10 g
UN Number UN2811
Merck Index 14,491
Solubility Information Soluble in dimethyl sulfoxide,ethanol,water,dimethyl formamide,acetone,chloroform,ethyl acetate and methanol.
Formula Weight 408.88
Percent Purity ≥97%
Physical Form Crystalline Powder
Chemical Name or Material Amlodipine
Hazard Category H301-H318-H373
Hazard Statement GHS H Statement
H301-H318-H373
Toxic if swallowed.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P305+P351+P338-P310-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 425-820-1
RTECSNumber US7968329
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only