Learn More
Amiodarone hydrochloride
A non-selective ion channel blocker; also an antiarrhythmic agent | CAS: 19774-82-4 | C25H30ClI2NO3 | 681.778 g/mol
Supplier: Thermo Scientific Chemicals J6045614
Description
Amiodarone induces an immediate influx of Ca2+ in Saccharomyces cerevisiae, followed by mitochondrial fragmentation and cell death.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAmiodarone induces an immediate influx of Ca2+ in Saccharomyces cerevisiae, followed by mitochondrial fragmentation and cell death.
Solubility
Soluble in chloroform, methanol.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
Specifications
| Amiodarone hydrochloride | |
| 156°C to 163°C | |
| C25H30ClI2NO3 | |
| 25 g | |
| amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron | |
| ITPDYQOUSLNIHG-UHFFFAOYSA-N | |
| (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride | |
| 441325 | |
| Powder |
| 19774-82-4 | |
| White | |
| MFCD00069204 | |
| 14,482 | |
| Soluble in chloroform,methanol. | |
| CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl | |
| 681.778 | |
| 681.77 |
Chemical Identifiers
| 19774-82-4 | |
| 681.778 | |
| ITPDYQOUSLNIHG-UHFFFAOYSA-N | |
| 441325 | |
| CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl |
| C25H30ClI2NO3 | |
| MFCD00069204 | |
| amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron | |
| (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride |
Safety and Handling
P201-P202-P260-P271-P281-P302+P352-P304+P340-P308+P313-P312-P363-P501c
H312+H332-H361d-H373
EINECSNumber : 243-293-2
RTECSNumber : OB1361000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only