missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 19774-82-4 |
|---|---|
| Molecular Formula | C25H29I2NO3·HCl |
| Molecular Weight (g/mol) | 681.77 |
| InChI Key | ITPDYQOUSLNIHG-UHFFFAOYSA-N |
| Synonym | amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron |
| PubChem CID | 441325 |
| IUPAC Name | (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride |
| SMILES | CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC457980050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 19774-82-4 | |
| 681.77 | |
| amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron | |
| (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride |
| C25H29I2NO3·HCl | |
| ITPDYQOUSLNIHG-UHFFFAOYSA-N | |
| 441325 | |
| CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl |
Specifications
| 19774-82-4 | |
| Authentic | |
| amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron | |
| ITPDYQOUSLNIHG-UHFFFAOYSA-N | |
| (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride | |
| 441325 | |
| ≥97.5% (HPLC) |
| 0.5% max. | |
| C25H29I2NO3·HCl | |
| Solubility in water: insoluble. | |
| CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl | |
| 681.77 | |
| 681.77 | |
| Amiodarone hydrochloride |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging the unborn child.
GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position
GHS Signal Word: Warning
EINECSNumber : 243-293-2
RUO – Research Use Only