missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amiodarone Hydrochloride 98.0+%, TCI America™
Supplier: TCI America A25305G
Specifications
| Amiodarone Hydrochloride | |
| 156°C | |
| C25H30ClI2NO3 | |
| 5 g | |
| ITPDYQOUSLNIHG-UHFFFAOYSA-N | |
| (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride | |
| 441325 | |
| ≥98.0% (HPLC,T) |
| 19774-82-4 | |
| White | |
| MFCD00069204 | |
| amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron | |
| CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl | |
| 681.778 | |
| 681.78 | |
| Crystalline Powder |
Chemical Identifiers
| 19774-82-4 | |
| 681.778 | |
| ITPDYQOUSLNIHG-UHFFFAOYSA-N | |
| 441325 | |
| CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl |
| C25H30ClI2NO3 | |
| MFCD00069204 | |
| amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron | |
| (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride |
Safety and Handling
RTECSNumber : OB1361000
TSCA : No
Recommended Storage : Refrigerator