missing translation for 'onlineSavingsMsg'
Learn More

Amiodarone Hydrochloride 98.0+%, TCI America™

Catalog No. A25301G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25301G 1 g
A25305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25301G Supplier TCI America Supplier No. A25301G

Chemical Identifiers

CAS 19774-82-4
Molecular Formula C25H30ClI2NO3
Molecular Weight (g/mol) 681.778
MDL Number MFCD00069204
InChI Key ITPDYQOUSLNIHG-UHFFFAOYSA-N
Synonym amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron
PubChem CID 441325
IUPAC Name (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride
SMILES CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl

Specifications

Melting Point 156°C
Color White
Quantity 1 g
Formula Weight 681.78
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Amiodarone Hydrochloride
RTECSNumber OB1361000
TSCA No
Recommended Storage Refrigerator