missing translation for 'onlineSavingsMsg'
Learn More

Aminopterin, Thermo Scientific™

Catalog No. ac468751000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468751000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC468751000 Supplier Thermo Scientific Chemicals Supplier No. 468751000
Only null left

Chemical Identifiers

CAS 54-62-6
Molecular Formula C19H20N8O5
Molecular Weight (g/mol) 440.42
MDL Number 00036692
InChI Key TVZGACDUOSZQKY-UHFFFAOYNA-N
IUPAC Name 2-[(4-{[(2,4-diaminopteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid
SMILES NC1=NC(N)=C2N=C(CNC3=CC=C(C=C3)C(=O)NC(CCC(O)=O)C(O)=O)C=NC2=N1

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color Light Yellow to Yellow/Brown
Melting Point 225°C
Merck Index 13-471
Solubility Soluble in DMF, DMSO
Quantity 100 mg
Chemical Name or Material Aminopterin
Hazard Category Skull and crossbones
Hazard Statement Health hazard
EINECSNumber 200-209-9
RTECSNumber MA1050000
Recommended Storage Normal conditions