missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Aminomethyl)phosphonic Acid, TCI America™
Supplier: TCI America A10725G
Specifications
| (Aminomethyl)phosphonic Acid | |
| White | |
| MFCD00008105 | |
| aminomethyl phosphonic acid, aminomethanephosphonic acid, amep, 1-aminomethylphosphonic acid, 1-aminomethylphosphonate, amino methane phosphoric acid, caswell no. 037c, 1-aminomethyl phosphonic acid, phosphonic acid, aminomethyl | |
| C(N)P(=O)(O)O | |
| 111.037 | |
| CHEBI:28812 | |
| ≥97.0% (T) |
| 1066-51-9 | |
| CH6NO3P | |
| 5 g | |
| MGRVRXRGTBOSHW-UHFFFAOYSA-N | |
| aminomethylphosphonic acid | |
| 14017 | |
| 111.04 | |
| Crystalline Powder |
Chemical Identifiers
| 1066-51-9 | |
| 111.037 | |
| MGRVRXRGTBOSHW-UHFFFAOYSA-N | |
| 14017 | |
| aminomethylphosphonic acid |
| CH6NO3P | |
| MFCD00008105 | |
| aminomethyl phosphonic acid, aminomethanephosphonic acid, amep, 1-aminomethylphosphonic acid, 1-aminomethylphosphonate, amino methane phosphoric acid, caswell no. 037c, 1-aminomethyl phosphonic acid, phosphonic acid, aminomethyl | |
| CHEBI:28812 | |
| C(N)P(=O)(O)O |
Safety and Handling
RTECSNumber : SZ6526000
TSCA : No