missing translation for 'onlineSavingsMsg'
Learn More

(Aminomethyl)phosphonic Acid, TCI America™

Catalog No. A10721G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10721G 1 g
A10725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10721G Supplier TCI America Supplier No. A10721G

Chemical Identifiers

CAS 1066-51-9
Molecular Formula CH6NO3P
Molecular Weight (g/mol) 111.037
MDL Number MFCD00008105
InChI Key MGRVRXRGTBOSHW-UHFFFAOYSA-N
Synonym aminomethyl phosphonic acid, aminomethanephosphonic acid, amep, 1-aminomethylphosphonic acid, 1-aminomethylphosphonate, amino methane phosphoric acid, caswell no. 037c, 1-aminomethyl phosphonic acid, phosphonic acid, aminomethyl
PubChem CID 14017
ChEBI CHEBI:28812
IUPAC Name aminomethylphosphonic acid
SMILES C(N)P(=O)(O)O

Specifications

Color White
Quantity 1 g
Formula Weight 111.04
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (Aminomethyl)phosphonic Acid
RTECSNumber SZ6526000
TSCA No