Learn More
(Aminomethyl)phosphonic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 291640010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (Aminomethyl)phosphonic acid | |
| 98.5 | |
| >300.0°C | |
| Authentic | |
| Glass bottle | |
| H2NCH2P(O)(OH)2 | |
| 1 g | |
| aminomethyl phosphonic acid, aminomethanephosphonic acid, amep, 1-aminomethylphosphonic acid, 1-aminomethylphosphonate, amino methane phosphoric acid, caswell no. 037c, 1-aminomethyl phosphonic acid, phosphonic acid, aminomethyl | |
| MGRVRXRGTBOSHW-UHFFFAOYSA-N | |
| aminomethylphosphonic acid | |
| 14017 | |
| 111.04 | |
| Fine Crystalline Powder |
| 1066-51-9 | |
| 100.0 | |
| White | |
| 99% | |
| CH6NO3P | |
| MFCD00008105 | |
| 01, 2596 | |
| Solubility in water: practically insoluble. Other solubilities: soluble in alcohols | |
| C(N)P(=O)(O)O | |
| 111.04 | |
| CHEBI:28812 | |
| 99% |
Chemical Identifiers
| 1066-51-9 | |
| 111.04 | |
| MGRVRXRGTBOSHW-UHFFFAOYSA-N | |
| 14017 | |
| aminomethylphosphonic acid |
| CH6NO3P | |
| MFCD00008105 | |
| aminomethyl phosphonic acid, aminomethanephosphonic acid, amep, 1-aminomethylphosphonic acid, 1-aminomethylphosphonate, amino methane phosphoric acid, caswell no. 037c, 1-aminomethyl phosphonic acid, phosphonic acid, aminomethyl | |
| CHEBI:28812 | |
| C(N)P(=O)(O)O |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only