missing translation for 'onlineSavingsMsg'
Learn More

Aminomethanesulfonic acid, 97%, Thermo Scientific™

Catalog No. AC155740500
Change view
Click to view available options
Quantity:
50g
500g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155740500 50g Glass bottle
AC155745000 500g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC155740500 Supplier Thermo Scientific Chemicals Supplier No. 155740500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 13881-91-9
Molecular Formula CH5NO3S
Molecular Weight (g/mol) 111.12
MDL Number MFCD00008124
InChI Key OBESRABRARNZJB-UHFFFAOYSA-N
Synonym methanesulfonic acid, amino, aminomethanesulphonic acid, aminomethyl sulfonic acid, aminomethylsulfonic acid, unii-ola224z482, methanesulfonic acid, 1-amino, aminomethanesulfonicacid, sodium aminomethanesulphonate, aminomethanesulfonic, aminomethanesulfoniccid
PubChem CID 83791
IUPAC Name aminomethanesulfonic acid
SMILES C(N)S(=O)(=O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula H2NCH2SO3H
Quantity 50g
Beilstein 01,583
Formula Weight 111.12
Percent Purity 97%
Chemical Name or Material Aminomethanesulfonic acid, 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 237-649-6
TSCA TSCA

RUO – Research Use Only