missing translation for 'onlineSavingsMsg'
Learn More

Aminomethanesulfonic acid, 97%

Catalog No. p-7027008
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2344122 100 g
AAB2344136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2344122 Supplier Thermo Scientific Chemicals Supplier No. B2344122

CAS: 13881-91-9 | CH5NO3S | 111.115 g/mol

Aminomethanesulfonic acid is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminomethanesulfonic acid is used as a pharmaceutical intermediates.

Solubility
Soluble in water. (17 g/L) at 17°C

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at room temperature.

Chemical Identifiers

CAS 13881-91-9
Molecular Formula CH5NO3S
Molecular Weight (g/mol) 111.115
MDL Number MFCD00008124
InChI Key OBESRABRARNZJB-UHFFFAOYSA-N
Synonym methanesulfonic acid, amino, aminomethanesulphonic acid, aminomethyl sulfonic acid, aminomethylsulfonic acid, unii-ola224z482, methanesulfonic acid, 1-amino, aminomethanesulfonicacid, sodium aminomethanesulphonate, aminomethanesulfonic, aminomethanesulfoniccid
PubChem CID 83791
IUPAC Name aminomethanesulfonic acid
SMILES C(N)S(=O)(=O)O

Specifications

Melting Point ∼180°C (decomposition)
pH 3.2
Quantity 100 g
UN Number UN2585
Beilstein 1811756
Solubility Information Soluble in water. (17g/L) at 17°C
Formula Weight 111.12
Percent Purity 97%
Chemical Name or Material Aminomethanesulfonic acid
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
EINECSNumber 237-649-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only