missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminomethanesulfonic Acid 97.0+%, TCI America™
$57.75 - $57.75
Chemical Identifiers
| CAS | 13881-91-9 |
|---|---|
| Molecular Formula | CH5NO3S |
| Molecular Weight (g/mol) | 111.115 |
| MDL Number | MFCD00008124 |
| InChI Key | OBESRABRARNZJB-UHFFFAOYSA-N |
| Synonym | methanesulfonic acid, amino, aminomethanesulphonic acid, aminomethyl sulfonic acid, aminomethylsulfonic acid, unii-ola224z482, methanesulfonic acid, 1-amino, aminomethanesulfonicacid, sodium aminomethanesulphonate, aminomethanesulfonic, aminomethanesulfoniccid |
| PubChem CID | 83791 |
| IUPAC Name | aminomethanesulfonic acid |
| SMILES | C(N)S(=O)(=O)O |
Chemical Identifiers
| 13881-91-9 | |
| 111.115 | |
| OBESRABRARNZJB-UHFFFAOYSA-N | |
| 83791 | |
| C(N)S(=O)(=O)O |
| CH5NO3S | |
| MFCD00008124 | |
| methanesulfonic acid, amino, aminomethanesulphonic acid, aminomethyl sulfonic acid, aminomethylsulfonic acid, unii-ola224z482, methanesulfonic acid, 1-amino, aminomethanesulfonicacid, sodium aminomethanesulphonate, aminomethanesulfonic, aminomethanesulfoniccid | |
| aminomethanesulfonic acid |
Specifications
| 13881-91-9 | |
| CH5NO3S | |
| 25 g | |
| methanesulfonic acid, amino, aminomethanesulphonic acid, aminomethyl sulfonic acid, aminomethylsulfonic acid, unii-ola224z482, methanesulfonic acid, 1-amino, aminomethanesulfonicacid, sodium aminomethanesulphonate, aminomethanesulfonic, aminomethanesulfoniccid | |
| C(N)S(=O)(=O)O | |
| 111.115 | |
| 111.12 | |
| Crystalline Powder |
| White | |
| MFCD00008124 | |
| 3261 | |
| OBESRABRARNZJB-UHFFFAOYSA-N | |
| aminomethanesulfonic acid | |
| 83791 | |
| ≥97.0% (T) | |
| Aminomethanesulfonic Acid |
Safety and Handling
EINECSNumber : (2)-3405
TSCA : Yes