missing translation for 'onlineSavingsMsg'
Learn More

Aminomalononitrile p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. A11195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11195G 5 g
A111925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11195G Supplier TCI America Supplier No. A11195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5098-14-6
Molecular Formula C10H11N3O3S
Molecular Weight (g/mol) 253.276
MDL Number MFCD00012848
InChI Key MEUWQVWJLLBVQI-UHFFFAOYSA-N
Synonym aminomalononitrile p-toluenesulfonate, 2-aminomalononitrile 4-methylbenzenesulfonate, aminomalononitrile p-toluenesulphonate, aminomalononitrile p-toluenesulfonic acid, aminomalononitrile tosylate, aminomalononitrile-p-toluene sulfonate, 2-aminomalononitrile 4-methylbenzenesulphonate, 2-aminomalononitrile p-toluenesulfonate, acmc-209krb, dicyanomethanaminium 4-methylbenzenesulfonate
PubChem CID 563049
IUPAC Name 2-aminopropanedinitrile;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.C(#N)C(C#N)N

Specifications

Melting Point 164°C
Quantity 5 g
UN Number 2811
Formula Weight 253.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Aminomalononitrile p-Toluenesulfonate
TSCA No
Recommended Storage Refrigerator