missing translation for 'onlineSavingsMsg'
Learn More

δ-Aminolevulinic acid hydrochloride, 99.7%, MP Biomedicals™

Catalog No. ICN10024790
Change view
Click to view available options
Quantity:
0.1 g
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10024790 500 mg
ICN10024780 0.1 g
ICN10024791 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10024790 Supplier MP Biomedicals Inc Supplier No. 0210024790

delta-Aminolevulinic acid hydrochloride is the universal precursor of tetrapyrroles, such as chlorophyll and heme

Chemical Identifiers

CAS 9-2-5451
Molecular Formula C5H10ClNO3
Molecular Weight (g/mol) 167.59
MDL Number MFCD00012869
InChI Key ZLHFONARZHCSET-UHFFFAOYSA-N
Synonym 5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride
PubChem CID 123608
IUPAC Name 5-amino-4-oxopentanoic acid;hydrochloride
SMILES C(CC(=O)O)C(=O)CN.Cl

Specifications

Color White
Assay Percent Range 0.997
Quantity 500 mg
Formula Weight 167.6
Physical Form Crystalline Powder
Chemical Name or Material δ-Aminolevulinic acid hydrochloride
EINECSNumber 226-679-5