Learn More
Aminoguanidine hydrogen carbonate, 98+%
CAS: 2582-30-1 | C2H8N4O3 | 136.111 g/mol
Supplier: Thermo Scientific Chemicals B246960E
Description
Aminoguanidine bicarbonate is an inhibitor of NOS (nitric oxide synthase). Aminoguanidine bicarbonate is used to study the effect of addition of polyamines to rat embryo cell cultures infected with adenovirus type 5.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAminoguanidine bicarbonate is an inhibitor of NOS (nitric oxide synthase). Aminoguanidine bicarbonate is used to study the effect of addition of polyamines to rat embryo cell cultures infected with adenovirus type 5.
Solubility
Soluble in water (3.3 mg/ml at 30°C), and water (2.7 mg/ml at 20°C).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| Aminoguanidine hydrogen carbonate | |
| ∼170°C (decomposition) | |
| C2H8N4O3 | |
| 2500 g | |
| 3569869 | |
| Soluble in water (3.3mg/ml at 30°C),and water (2.7mg/ml at 20°C). | |
| C(=NN)(N)N.C(=O)(O)O | |
| 136.111 | |
| 136.11 |
| 2582-30-1 | |
| 1.6 | |
| MFCD00012949 | |
| UN3077 | |
| aminoguanidine bicarbonate, aminoguanidine hydrogen carbonate, aminoguanidinium bicarbonate, aminoguanidine hydrocarbonate, aminoguanidine carbonate 1:1, aminoguanidium hydrogen carbonate, n1-aminoguanidine carbonate 1:1, aminoguanidine hydrogencarbonate, aminoguanidinebicarbonate, hydrazinecarboximidamide carbonate | |
| OTXHZHQQWQTQMW-UHFFFAOYSA-N | |
| 2-aminoguanidine;carbonic acid | |
| 164944 | |
| ≥98% |
Chemical Identifiers
| 2582-30-1 | |
| 136.111 | |
| OTXHZHQQWQTQMW-UHFFFAOYSA-N | |
| 164944 | |
| C(=NN)(N)N.C(=O)(O)O |
| C2H8N4O3 | |
| MFCD00012949 | |
| aminoguanidine bicarbonate, aminoguanidine hydrogen carbonate, aminoguanidinium bicarbonate, aminoguanidine hydrocarbonate, aminoguanidine carbonate 1:1, aminoguanidium hydrogen carbonate, n1-aminoguanidine carbonate 1:1, aminoguanidine hydrogencarbonate, aminoguanidinebicarbonate, hydrazinecarboximidamide carbonate | |
| 2-aminoguanidine;carbonic acid |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H360
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 219-956-7
RTECSNumber : FG1772000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only