missing translation for 'onlineSavingsMsg'
Learn More

Aminoguanidine Hydrochloride 98.0+%, TCI America™

Catalog No. A112925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A112925G 25 g
A1129100G 100 g
A1129500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A112925G Supplier TCI America Supplier No. A112925G

Chemical Identifiers

CAS 1937-19-5
Molecular Formula CH7ClN4
Molecular Weight (g/mol) 110.545
MDL Number MFCD00039074
InChI Key UBDZFAGVPPMTIT-UHFFFAOYSA-N
Synonym aminoguanidine hydrochloride, pimagedine hcl, guanylhydrazine hydrochloride, hydrazinecarboximidamide hydrochloride, aminoguanidinehydrochloride, aminoguanidine hcl, pimagedine hydrochloride, aminoguanidine monohydrochloride, hydrazinecarboximidamide, hydrochloride
PubChem CID 2734687
IUPAC Name 2-aminoguanidine;hydrochloride
SMILES C(=NN)(N)N.Cl

Specifications

Melting Point 165°C
Color White
Quantity 25 g
Formula Weight 110.55
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Aminoguanidine Hydrochloride
RTECSNumber ME8430000
TSCA Yes