missing translation for 'onlineSavingsMsg'
Learn More

Aminoguanidine Bicarbonate 99.0+%, TCI America™

Catalog No. A030725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A030725G 25 g
A0307500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A030725G Supplier TCI America Supplier No. A030725G

Chemical Identifiers

CAS 2582-30-1
Molecular Formula C2H8N4O3
Molecular Weight (g/mol) 136.111
MDL Number MFCD00012949
InChI Key OTXHZHQQWQTQMW-UHFFFAOYSA-N
Synonym aminoguanidine bicarbonate, aminoguanidine hydrogen carbonate, aminoguanidinium bicarbonate, aminoguanidine hydrocarbonate, aminoguanidine carbonate 1:1, aminoguanidium hydrogen carbonate, n1-aminoguanidine carbonate 1:1, aminoguanidine hydrogencarbonate, aminoguanidinebicarbonate, hydrazinecarboximidamide carbonate
PubChem CID 164944
IUPAC Name 2-aminoguanidine;carbonic acid
SMILES C(=NN)(N)N.C(=O)(O)O

Specifications

Melting Point 162°C
Color White
Quantity 25 g
Formula Weight 136.11
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Aminoguanidine Bicarbonate
EINECSNumber (2)-2884
RTECSNumber FG1772000
TSCA Yes