missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminoferrocene 96.0+%, TCI America™
$186.41 - $753.08
Chemical Identifiers
| CAS | 1273-82-1 |
|---|---|
| Molecular Formula | C10H11FeN |
| MDL Number | MFCD03844760 |
| Synonym | Ferrocenylamine |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A1601100MG
|
TCI America
A1601100MG |
100 mg |
Each for $186.41
|
|
|||||
|
A16011G
|
TCI America
A16011G |
1 g |
Each for $753.08
|
|
|||||
Chemical Identifiers
| 1273-82-1 | |
| C10H11FeN | |
| MFCD03844760 | |
| Ferrocenylamine |
Specifications
| 1273-82-1 | |
| MFCD03844760 | |
| Ferrocenylamine | |
| ≥96.0% (GC,T) | |
| Aminoferrocene |
| C10H11FeN | |
| 100 mg | |
| 201.05 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator